Order Entry
Italy
ContactUsLinkComponent
 
Anti-MFN1 Mouse Monoclonal Antibody [clone: 11E9-1H12]
 
undefined
Anti-MFN1 Mouse Monoclonal Antibody [clone: 11E9-1H12]

 

  • Catalog No:
  • NOVUNBP1-71775SS
  • NOVUNBP1-71775
  • Antigen symbol:
  • MFN1
  • MFN1
  • Antigen name:
  • Mitofusin 1
  • Mitofusin 1
  • Antigen synonyms:
  • hfzo1|hfzo2
  • hfzo1|hfzo2
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • 11E9-1H12
  • 11E9-1H12
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 55669
  • 55669
  • Isotype:
  • IgG2b kappa
  • IgG2b kappa
  • Western blot:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

 

 

Frequently Bought Together