Anti-TP53INP2 Rabbit Polyclonal Antibody (DyLight® 650)
Codice catalogo NOVUNBP1-71704C
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Tumor Protein 53-inducible Nuclear Protein 2
- Antigen symbol:TP53INP2
- Clonality:Polyclonal
- Conjugation:DyLight® 650
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human,Mouse
- Gene ID:58476
- Antigen synonyms:PINH|DOR|C20orf110
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: TP53INP2 (tumor protein p53 inducible nuclear protein 2)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse