Order Entry
Italy
ContactUsLinkComponent
Anti-TP53INP2 Rabbit Polyclonal Antibody
Anti-TP53INP2 Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals
Certificates
undefined
Anti-TP53INP2 Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals

Specifications

  • Catalog No:
  • NOVUNBP1-71704SS
  • NOVUNBP1-71704
  • Antigen symbol:
  • TP53INP2
  • TP53INP2
  • Antigen name:
  • Tumor Protein 53-inducible Nuclear Protein 2
  • Tumor Protein 53-inducible Nuclear Protein 2
  • Antigen synonyms:
  • PINH|DOR|C20orf110
  • PINH|DOR|C20orf110
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • 58476
  • 58476
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Related Information

Frequently Bought Together