Order Entry
Italy
ContactUsLinkComponent
Anti-mtTFA Mouse Monoclonal Antibody (DyLight® 650) [clone: 18G102B2E11]
Anti-mtTFA Mouse Monoclonal Antibody (DyLight® 650) [clone: 18G102B2E11]
Catalog # NOVUNBP1-71648C
Supplier:  Novus Biologicals
Anti-mtTFA Mouse Monoclonal Antibody (DyLight® 650) [clone: 18G102B2E11]
Catalog # NOVUNBP1-71648C
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Transcription Factor A, Mitochondrial
  • Antigen symbol:
    TFAM
  • Clonality:
    Monoclonal
  • Clone:
    18G102B2E11
  • Conjugation:
    DyLight® 650
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG2b kappa
  • Reactivity:
    Human,
    Rat
  • Western blot:
    Yes
  • Gene ID:
    7019
  • Antigen synonyms:
    mitochondrial transcription factor A|mtTFA|TCF-6|TCF6L1|mitochondrial transcription factor 1|Transcription factor 6-like 2 (mitochondrial transcription factor)|TCF6L2|transcription factor 6|TCF6L3|TCF6|MtTF1
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µL
  • Pk:
    0,1 mL

Specifications

About this item

Type: Primary
Antigen: mtTFA
Clonality: monoclonal
Clone: 18G102B2E11
Conjugation: DyLight® 650
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human, Rat