Order Entry
Italy
ContactUsLinkComponent
Anti-TGOLN2 Rabbit Polyclonal Antibody (DyLight® 488)
Anti-TGOLN2 Rabbit Polyclonal Antibody (DyLight® 488)
  NOVUNBP1-49643G
Anti-TGOLN2 Rabbit Polyclonal Antibody (DyLight® 488)
  NOVUNBP1-49643G

 

  • Antibody type:
    Primary
  • Antigen name:
    Trans-Golgi Network Integral Membrane Protein 2
  • Antigen symbol:
    TGOLN2
  • Clonality:
    Polyclonal
  • Conjugation:
    DyLight® 488
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Human
  • Gene ID:
    10618
  • Antigen synonyms:
    TGN38|trans-golgi network protein 2|TGN51|TGN48|TTGN2|TGN46
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µL
  • Pk:
    0,1 mL

 

 

Type: Primary
Antigen: TGOLN2 (trans-golgi network protein 2)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 488
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human