Anti-KDM3A Rabbit Polyclonal Antibody (DyLight® 650)
Catalog # NOVUNBP1-49601C
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Lysine-specific demethylase 3A
- Antigen symbol:JMJD1A
- Clonality:Polyclonal
- Conjugation:DyLight® 650
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Gene ID:55818
- Antigen synonyms:JHDM2A|TSGA|JHMD2A|JMJD1A|JMJD1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Specifications
About this item
Rabbit Polyclonal JMJD1A Antibody [DyLight 650]. Tested Applications: Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Human, Mouse.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:50
Type: Primary
Antigen: JMJD1A
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human