Anti-C12orf5 Rabbit Polyclonal Antibody (DyLight® 488)
Catalog # NOVUNBP1-49534G
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:TP53 Induced Glycolysis Regulatory Phosphatase
- Antigen symbol:TIGAR
- Clonality:Polyclonal
- Conjugation:DyLight® 488
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Gene ID:57103
- Antigen synonyms:Chromosome 12 Open Reading Fame 5|C12orf5|FR2BP
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: C12orf5 (chromosome 12 open reading frame 5)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 488
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse