Anti-ApoE4 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 4E4]
Catalog # NOVUNBP1-49529H
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Apolipoprotein E4
- Antigen symbol:APOE4
- Clonality:Monoclonal
- Clone:4E4
- Conjugation:HRP (Horseradish Peroxidase)
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Gene ID:348
- Antigen synonyms:ApoE4
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: ApoE4
Clonality: monoclonal
Clone: 40000
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human