Anti-EGLN3 Mouse Monoclonal Antibody (DyLight® 405) [clone: EG188e/d5]
Catalog # NOVUNBP1-30440V
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:egl-9 family hypoxia-inducible factor 3
- Antigen symbol:EGLN3
- Clonality:Monoclonal
- Clone:EG188e/d5
- Conjugation:DyLight® 405
- Host:Mouse
- Isotype:IgG1
- Western blot:Yes
- Gene ID:112399
- Antigen synonyms:PHD3|HIFPH3|HIFP4H3
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:200 μL
- Pk:0,2 mL
Specifications
About this item
Type: Primary
Antigen: EGLN3 (egl-9 family hypoxia-inducible factor 3)
Clonality: monoclonal
Clone: EG188e/d5
Conjugation: DyLight® 405
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: