Anti-HIF1A Mouse Monoclonal Antibody (DyLight® 550) [clone: 162c]
NOVUNBP1-30333R
- Antibody type:Primary
- Antigen name:Factor Inhibiting HIF-1
- Clonality:Monoclonal
- Clone:162c
- Conjugation:DyLight® 550
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Gene ID:55662
- Antigen synonyms:FIH1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Type: Primary
Antigen: HIF1A (hypoxia inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor))
Clonality: monoclonal
Clone: 162c
Conjugation: DyLight® 550
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human