Anti-EGLN1 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 366G/76/3]
Catalog # NOVUNBP1-30328F
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:egl-9 family hypoxia-inducible factor 1
- Antigen symbol:EGLN1
- Clonality:Monoclonal
- Clone:366G/76/3
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Gene ID:54583
- Antigen synonyms:HPH-2|HALAH|PHD2|HIFPH2|HIF-PH2|ECYT3|C1orf12|ZMYND6|HPH2|SM20
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: EGLN1 (egl-9 family hypoxia-inducible factor 1)
Clonality: monoclonal
Clone: 366G/76/3
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human