Anti-EGLN1 Mouse monoclonal antibody unconjugated [clone: 366G/76/3]
Supplier: Novus Biologicals
Certificates
About this item
Type: Primary
Antigen: EGLN1 (egl-9 family hypoxia-inducible factor 1)
Clonality: monoclonal
Clone: 366G/76/3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Specifications
- Catalog No:
- NOVUNBP1-30328
- NOVUNBP1-30328SS
- Antigen symbol:
- EGLN1
- EGLN1
- Antigen name:
- egl-9 family hypoxia-inducible factor 1
- egl-9 family hypoxia-inducible factor 1
- Antigen synonyms:
- HPH-2|HALAH|PHD2|HIFPH2|HIF-PH2|ECYT3|C1orf12|ZMYND6|HPH2|SM20
- HPH-2|HALAH|PHD2|HIFPH2|HIF-PH2|ECYT3|C1orf12|ZMYND6|HPH2|SM20
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- 366G/76/3
- 366G/76/3
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 54583
- 54583
- Isotype:
- IgG1
- IgG1
- Western blot:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Specifications
Frequently Bought Together