Order Entry
Italy
ContactUsLinkComponent
Anti-HIST3H3 Mouse monoclonal antibody unconjugated [clone: 6F12-H4]
Anti-HIST3H3 Mouse monoclonal antibody unconjugated [clone: 6F12-H4]
Supplier:  Novus Biologicals
Certificates
Anti-HIST3H3 Mouse monoclonal antibody unconjugated [clone: 6F12-H4]
Supplier:  Novus Biologicals

Specifications

  • Catalog No:
  • NOVUNBP1-30141
  • NOVUNBP1-30141SS
  • Antigen symbol:
  • H3
  • H3
  • Antigen name:
  • Histone H3
  • Histone H3
  • Antigen synonyms:
  • H3FA|HIST1H3J|H3FJ|HIST1H3E|HIST1H3G|HIST1H3B|H3FI|HIST1H3D|HIST1H3C|HIST1H3I|HIST1H3F|H3T|HIST3H3|H3FH|MGC126886|H3FD|H3FF|HIST1H3H|H3FC|H3FB|H3FT|H3FL|H3FK|HIST1H3A
  • H3FA|HIST1H3J|H3FJ|HIST1H3E|HIST1H3G|HIST1H3B|H3FI|HIST1H3D|HIST1H3C|HIST1H3I|HIST1H3F|H3T|HIST3H3|H3FH|MGC126886|H3FD|H3FF|HIST1H3H|H3FC|H3FB|H3FT|H3FL|H3FK|HIST1H3A
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • 6F12-H4
  • 6F12-H4
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 126961
  • 126961
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Western blot:
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • ChIP:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Related Information

Frequently Bought Together