Anti-HIST4H4 Mouse Monoclonal Antibody (DyLight® 488) [clone: 5E10-D8]
Codice catalogo NOVUNBP1-30091G
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Histone cluster 4 H4
- Antigen symbol:HIST4H4
- Clonality:Monoclonal
- Clone:5E10-D8
- Conjugation:DyLight® 488
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Western blot:Yes
- ChIP:Yes
- Gene ID:121504
- Antigen synonyms:H4FJ|HIST1H4H|H4/I|H4/M|H4FI|H4/K|H4FA|H4/G|HIST2H4B|H4F2|H4/O|HIST1H4L|H4/D|H4FC|H4/E|H4/B|H4FE|HIST1H4J|H4FH|H4FM|H4FO|H4FB|HIST1H4K|H4/N|H4/p|H4/J|HIST2H4|H4/C|HIST1H4F|H4FG|HIST1H4I|H4/A|H4FK|H4/H|H4FN|H4FD|HIST2H4A
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: HIST4H4 (histone cluster 4, H4)
Clonality: monoclonal
Clone: 5E10-D8
Conjugation: DyLight® 488
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: