Anti-HIST4H4 Mouse Monoclonal Antibody (DyLight® 680) [clone: 5E10-D8]
Catalog # NOVUNBP1-30091FR
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:histone cluster 4, H4
- Antigen symbol:HIST4H4
- Clonality:Monoclonal
- Clone:5E10-D8
- Conjugation:DyLight® 680
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Western blot:Yes
- ChIP:Yes
- Gene ID:121504
- Antigen synonyms:H4/p
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 Tests
- Pk:100 Tests
Specifications
About this item
Type: Primary
Antigen: HIST4H4 (histone cluster 4, H4)
Clonality: monoclonal
Clone: 5E10-D8
Conjugation: DyLight® 680
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: