Anti-BIRC3 Rabbit Polyclonal Antibody (DyLight® 488)
Catalog # NOVUNBP1-27972G
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:baculoviral IAP repeat containing 3
- Antigen symbol:cIAP2
- Clonality:Polyclonal
- Conjugation:DyLight® 488
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Gene ID:330
- Antigen synonyms:MIHC|MALT2|HAIP1|API2|AIP1|RNF49|CIAP2|HIAP1|c-IAP2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: BIRC3 (baculoviral IAP repeat containing 3)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 488
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human