Anti-KDM6B Rabbit Polyclonal Antibody (DyLight® 488)
Catalog # NOVUNBP1-06640G
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Jumonji Domain-Containing protein 3
- Antigen symbol:JMJD3
- Clonality:Polyclonal
- Conjugation:DyLight® 488
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Gene ID:23135
- Antigen synonyms:JmjC domain-containing protein 3|KIAA0346|Lysine demethylase 6B|Jumonji domain-containing protein 3|KDM6B|Lysine-specific demethylase 6B|JMJD3
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: KDM6B (lysine (K)-specific demethylase 6B)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 488
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse