Anti-ZIC1 Rabbit Polyclonal Antibody (DyLight® 550)
Catalog # NOVUNB600-488R
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Zic Family member 1
- Antigen symbol:ZIC1
- Clonality:Polyclonal
- Conjugation:DyLight® 550
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Gene ID:7545
- Antigen synonyms:Zic1 encodes a zinc finger protein expressed in the developing or matured central nervous system in a highly restricted manner. Zic is expressed in granule cells that make synaptic contact with Purkinje cells. Zic1 is a gene critical to cerebellar pattern formation. The expression of Zic genes is first detected at gastrulation and at neurulation|becoming restricted to the dorsal neural ectoderm and the dorsal paraxial mesoderm. The Zic1 gene has been mapped to chromosome 9 in mouse. The 5' flanking region of the Zic1 gene contains a region-specific enhancer determined to be essential in in-vivo and in-vitro deletion analysis. The temporal profile of mRNA expression differs for each of the Zic gene products. The Drosophila odd-paired gene is highly homologous to the Zic gene family. Zic2 and Zic3 are highly similar genes. Zic2 is essential for the formation of the brain and Zic-3 is important for right and left axis formation.
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: ZIC1 (Zic family member 1)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 550
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse