Order Entry
Italy
ContactUsLinkComponent
 
Anti-CASP3 Mouse monoclonal antibody unconjugated [clone: CPP32 4-1-18]
 
undefined
Anti-CASP3 Mouse monoclonal antibody unconjugated [clone: CPP32 4-1-18]

 

  • Catalog No:
  • NOVUNB500-210
  • NOVUNB500-210SS
  • Antigen symbol:
  • CASP3
  • CASP3
  • Antigen name:
  • Caspase 3
  • Caspase 3
  • Antigen synonyms:
  • caspase-3|procaspase3|caspase 3|protein Yama|Yama|cysteine protease CPP32|PARP cleavage protease|CPP32|apoptosis-related cysteine protease|CPP32B|SCA-1|apopain|SREBP cleavage activity 1
  • caspase-3|procaspase3|caspase 3|protein Yama|Yama|cysteine protease CPP32|PARP cleavage protease|CPP32|apoptosis-related cysteine protease|CPP32B|SCA-1|apopain|SREBP cleavage activity 1
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • CPP32 4-1-18
  • CPP32 4-1-18
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 836
  • 836
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa
  • Western blot:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

 

 

Frequently Bought Together