Order Entry
Italy
Orders LinkContactUsLinkComponent
Anti-POT1 Rabbit Polyclonal Antibody

Anti-POT1 Rabbit Polyclonal Antibody

Supplier:   Novus Biologicals
Anti-POT1 Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals
Certificates
Anti-POT1 Rabbit Polyclonal Antibody

2 Options Available Below

Product Details & Documents

The POT1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to POT1. This antibody reacts with human. The POT1 Antibody has been validated for the following applications: Western Blot.

There is 1 scientific publication using this antibody.

Type: Primary
Antigen: POT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Documents

Specifications for Product Family

Specifications

  • Catalog No:
  • NOVUNB500-176
  • NOVUNB500-176SS
  • Antigen symbol:
  • POT1
  • POT1
  • Antigen name:
  • Protection of telomeres 1
  • Protection of telomeres 1
  • Antigen synonyms:
  • POT 1|POT-1|Protection of telomeres 1
  • POT 1|POT-1|Protection of telomeres 1
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • 25913
  • 25913
  • Western blot:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Product Family Options

Product Information

  • Size
    Availability
    Price
  • 100 µL
    Log in for availability
    Log in for availability
    1 * 0,1 mL
    1 * 0,1 mL - 658,00 €
    658,00 €
    1 * 0,1 mL
       658,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    NOVUNB500-176
    Antigen symbol
    POT1
    Antigen name
    Protection of telomeres 1
    Antigen synonyms
    POT 1|POT-1|Protection of telomeres 1
    Antibody type
    Primary
    Clonality
    Polyclonal
    Conjugation
    Unconjugated
    Reactivity
    Human
    Host
    Rabbit
    Gene ID
    25913
    Western blot
    Yes
    Storage temperature
    Cool
    Shipping temperature
    Ice
  • 25 µL
    Log in for availability
    Log in for availability
    1 * 25 µl
    1 * 25 µl - 284,00 €
    284,00 €
    1 * 25 µl
       284,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    NOVUNB500-176SS
    Antigen symbol
    POT1
    Antigen name
    Protection of telomeres 1
    Antigen synonyms
    POT 1|POT-1|Protection of telomeres 1
    Antibody type
    Primary
    Clonality
    Polyclonal
    Conjugation
    Unconjugated
    Reactivity
    Human
    Host
    Rabbit
    Gene ID
    25913
    Western blot
    Yes
    Storage temperature
    Cool
    Shipping temperature
    Ice