Anti-MKI67 Rabbit Polyclonal Antibody (FITC (Fluorescein))
NOVUNB500-170F
- Antibody type:Primary
- Antigen name:Marker of proliferation Ki-67
- Antigen symbol:MKI67
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Gene ID:4288
- Antigen synonyms:MIB-1|MIB-|KIA|PPP1R105
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μL
- Pk:0,1 mL
Type: Primary
Antigen: MKI67 (marker of proliferation Ki-67)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: