Anti-Neurokinin B Rabbit Polyclonal Antibody
Supplier: Novus Biologicals
Certificates
About this item
Type: Primary
Antigen: Neurokinin B
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Specifications
- Catalog No:
- NOVUNB300-201SS
- NOVUNB300-201
- Antigen symbol:
- NKB
- NKB
- Antigen name:
- Neurokinin B
- Neurokinin B
- Antigen synonyms:
- NKB|PRO1155|ZNEUROK1|NKNB|HH10
- NKB|PRO1155|ZNEUROK1|NKNB|HH10
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 6866
- 6866
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Specifications
Frequently Bought Together