Order Entry
Italy
ContactUsLinkComponent
Anti-HIST3H3 Rabbit Polyclonal Antibody (Biotin)
Anti-HIST3H3 Rabbit Polyclonal Antibody (Biotin)
  NOVUNB21-1012B
Anti-HIST3H3 Rabbit Polyclonal Antibody (Biotin)
  NOVUNB21-1012B

 

  • Antibody type:
    Primary
  • Antigen name:
    Histone Cluster 2, H3c
  • Antigen symbol:
    HIST2H3C
  • Clonality:
    Polyclonal
  • Conjugation:
    Biotin
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Isotype:
  • Reactivity:
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Gene ID:
    126961
  • Antigen synonyms:
    H3.3B|family 3A|H3R2me1|MGC87782|H3.3AH3F3H3F3B|histone H3.3|MGC87783|H3 histone
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    50 µg
  • Pk:
    50 µG

 

 

Type: Primary
Antigen: HIST3H3 (histone cluster 3, H3)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity: