Order Entry
Italy
ContactUsLinkComponent
Anti-TERF1 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 57-6]
Anti-TERF1 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 57-6]
Catalog # NOVUNB110-68281F
Supplier:  Novus Biologicals
Anti-TERF1 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 57-6]
Catalog # NOVUNB110-68281F
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Telomeric Repeat Binding Factor 1
  • Antigen symbol:
    TERF1
  • Clonality:
    Monoclonal
  • Clone:
    57-6
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Western blot:
    Yes
  • Gene ID:
    7013
  • Antigen synonyms:
    TRF1|hTRF1-AS|TRBF1|PIN2|t-TRF1|TRF
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 μL
  • Pk:
    0,1 mL

Specifications

About this item

Type: Primary
Antigen: TERF1 (telomeric repeat binding factor (NIMA-interacting) 1)
Clonality: monoclonal
Clone: 57-6
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: