Anti-PLIN5 Rabbit Polyclonal Antibody (DyLight® 405)
Catalog # NOVUNB110-60511V
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:perilipin 5
- Antigen symbol:PLIN5
- Clonality:Polyclonal
- Conjugation:DyLight® 405
- Host:Rabbit
- Reactivity:Human,Mouse
- Western blot:Yes
- Gene ID:440503
- Antigen synonyms:MLDP|LSDA5|LSDP5|OXPAT
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: PLIN5 (perilipin 5)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 405
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse