Anti-PLIN5 Rabbit Polyclonal Antibody
Type: Primary
Antigen: PLIN5 (perilipin 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
- Catalog No:
- NOVUNB110-60509SS
- NOVUNB110-60509
- Antigen symbol:
- OXPAT
- OXPAT
- Antigen name:
- Perilipin-5
- Perilipin-5
- Antigen synonyms:
- MLDP|LSDA5|LSDP5|OXPAT
- MLDP|LSDA5|LSDP5|OXPAT
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 440503
- 440503
- Western blot:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Frequently Bought Together