Order Entry
Italy
ContactUsLinkComponent
Anti-ATP13A2 Rabbit Polyclonal Antibody
Anti-ATP13A2 Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals
Certificates
undefined
Anti-ATP13A2 Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals

Specifications

  • Catalog No:
  • NOVUNB110-41486SS
  • NOVUNB110-41486
  • Antigen symbol:
  • ATP13A2
  • ATP13A2
  • Antigen name:
  • ATPase type 13A2
  • ATPase type 13A2
  • Antigen synonyms:
  • ATP13A2
  • ATP13A2
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • 23400
  • 23400
  • Western blot:
  • Yes
  • Yes
  • Storage buffer:
  • Tris-Glycine and 0,15M NaCl
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Related Information

Frequently Bought Together