Order Entry
Italy
ContactUsLinkComponent
Anti-SLIRP Rabbit Polyclonal Antibody (DyLight® 488)
Anti-SLIRP Rabbit Polyclonal Antibody (DyLight® 488)
Codice catalogo NOVUNB110-37258G
Supplier:  Novus Biologicals
undefined
Anti-SLIRP Rabbit Polyclonal Antibody (DyLight® 488)
Codice catalogo NOVUNB110-37258G
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    SRA Stem-Loop Interacting RNA Binding Protein
  • Clonality:
    Polyclonal
  • Conjugation:
    DyLight® 488
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Gene ID:
    81892
  • Antigen synonyms:
    C14orf156|DC50|PD04872
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µL
  • Pk:
    0,1 mL

Specifications

About this item

Type: Primary
Antigen: SLIRP (SRA stem-loop interacting RNA binding protein)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 488
Epitope:
Host: Rabbit
Isotype: none
Reactivity: