Anti-PTPRC Rat Monoclonal Antibody (DyLight® 488) [clone: 30-F11]
Catalog # NOVUNB100-77417G
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Protein tyrosine phosphatase receptor type C
- Antigen symbol:PTPRC
- Clonality:Monoclonal
- Clone:30-F11
- Conjugation:DyLight® 488
- Flow cytometry:Yes
- Host:Rat
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2b kappa
- Reactivity:Mouse
- Gene ID:5788
- Antigen synonyms:CD45R|B220|CD45|GP180|T200|L-CA|LY5|receptor type C|LCA|protein tyrosine phosphatase
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: PTPRC (protein tyrosine phosphatase, receptor type, C)
Clonality: monoclonal
Clone: 30-F11
Conjugation: DyLight® 488
Epitope:
Host: Rat
Isotype: IgG2b Kappa
Reactivity: Mouse