Order Entry
Italy
ContactUsLinkComponent
Anti-rRNA Mouse Monoclonal Antibody (DyLight® 680) [clone: Y10b]
Anti-rRNA Mouse Monoclonal Antibody (DyLight® 680) [clone: Y10b]
Catalog # NOVUNB100-662FR
Supplier:  Novus Biologicals
Anti-rRNA Mouse Monoclonal Antibody (DyLight® 680) [clone: Y10b]
Catalog # NOVUNB100-662FR
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    rRNA
  • Clonality:
    Monoclonal
  • Clone:
    Y10b
  • Conjugation:
    DyLight® 680
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG3 kappa
  • Reactivity:
    Multiple
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 Tests
  • Pk:
    100 Tests

Specifications

About this item

Type: Primary
Antigen: rRNA
Clonality: monoclonal
Clone: Y10b
Conjugation: DyLight® 680
Epitope:
Host: Mouse
Isotype: IgG3 Kappa
Reactivity: Pan