Anti-Importin-9 Rabbit Polyclonal Antibody
Catalog # NOVUNB100-56489
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:importin 9
- Antigen symbol:IPO9
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Gene ID:55705
- Antigen synonyms:Imp9
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: Importin-9
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: