Anti-XAF1 Rabbit Polyclonal Antibody
Catalog # NOVUNB100-56355
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:XIAP Associated Factor 1
- Antigen symbol:XAF1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Gene ID:54739
- Antigen synonyms:XIAPAF1|HSXIAPAF1|BIRC4-binding protein|BIRC4BP|XIAP-associated factor 1|XAF-1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: XAF1 (XIAP associated factor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse