Anti-Cre Rabbit Polyclonal Antibody
Supplier: Novus Biologicals
Certificates
About this item
The Cre Antibody from Novus Biologicals is a rabbit polyclonal antibody to Cre. This antibody reacts with mouse, cre/lox system. The Cre Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:1000-1:5000
Type: Primary
Antigen: Cre
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Cre/lox System
Specifications
- Catalog No:
- NOVUNB100-56133
- NOVUNB100-56133SS
- Antigen symbol:
- Cre
- Cre
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Cre/lox system
- Cre/lox system
- Host:
- Rabbit
- Rabbit
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Storage buffer:
- PBS and 30% Glycerol
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Specifications
Frequently Bought Together