Order Entry
Italy
ContactUsLinkComponent
Anti-NOD2 Mouse Monoclonal Antibody [clone: 2D9]
Anti-NOD2 Mouse Monoclonal Antibody [clone: 2D9]
Supplier:  Novus Biologicals
Certificates
undefined
Anti-NOD2 Mouse Monoclonal Antibody [clone: 2D9]
Supplier:  Novus Biologicals

Specifications

  • Catalog No:
  • NOVUNB100-524
  • NOVUNB100-524SS
  • Antigen symbol:
  • NOD2
  • NOD2
  • Antigen name:
  • Nucleotide binding oligomerization domain containing 2
  • Nucleotide binding oligomerization domain containing 2
  • Antigen synonyms:
  • NLRC2|PSORAS1|NOD2|CARD15|CD|IBD1|ACUG|NOD2B|CLR16.3|BLAU
  • NLRC2|PSORAS1|NOD2|CARD15|CD|IBD1|ACUG|NOD2B|CLR16.3|BLAU
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • 2D9
  • 2D9
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 64127
  • 64127
  • Isotype:
  • IgG1
  • IgG1
  • Western blot:
  • Yes
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Related Information

Frequently Bought Together