Order Entry
Italy
ContactUsLinkComponent
 
Anti-NSP 5 alpha 3 alpha Rabbit Polyclonal Antibody (DyLight® 550)
  NOVUNB100-518R
Anti-NSP 5 alpha 3 alpha Rabbit Polyclonal Antibody (DyLight® 550)
  NOVUNB100-518R

 

  • Antibody type:
    Primary
  • Antigen symbol:
    NSP 5 alpha 3 alpha
  • Clonality:
    Polyclonal
  • Conjugation:
    DyLight® 550
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Gene ID:
    92521
  • Antigen synonyms:
    HCMOGT1|CYTSB|NSP|HCMOGT-1
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µL
  • Pk:
    0,1 mL

 

 

Type: Primary
Antigen: NSP 5 alpha 3 alpha
Clonality: polyclonal
Clone:
Conjugation: DyLight® 550
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human