Anti-BP1 Rabbit Polyclonal Antibody
Supplier: Novus Biologicals
Certificates
About this item
The BP1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to BP1. This antibody reacts with human. The BP1 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
There is 1 scientific publication using this antibody.
Type: Primary
Antigen: BP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- NOVUNB100-481SS
- NOVUNB100-481
- Antigen symbol:
- EIF4EBP1
- EIF4EBP1
- Antigen name:
- Eukaryotic translation initiation factor 4E binding protein 1
- Eukaryotic translation initiation factor 4E binding protein 1
- Antigen synonyms:
- DLX8|DLX7|DLX9|BP1
- DLX8|DLX7|DLX9|BP1
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 1748
- 1748
- Western blot:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- Storage buffer:
- Tris-Citrate/Phosphate (pH 7,0 - 8,0)
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Specifications
Frequently Bought Together