Order Entry
Italy
ContactUsLinkComponent
Anti-ATM Mouse monoclonal antibody unconjugated [clone: 10H11.E12]
Anti-ATM Mouse monoclonal antibody unconjugated [clone: 10H11.E12]
Supplier:  Novus Biologicals
Certificates
Anti-ATM Mouse monoclonal antibody unconjugated [clone: 10H11.E12]
Supplier:  Novus Biologicals

Specifications

  • Catalog No:
  • NOVUNB100-306
  • NOVUNB100-306SS
  • Antigen symbol:
  • ATM
  • ATM
  • Antigen name:
  • Ataxia Telangiectasia mutated
  • Ataxia Telangiectasia mutated
  • Antigen synonyms:
  • ATM Protein Kinase pS1981
  • ATM Protein Kinase pS1981
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • 10H11.E12
  • 10H11.E12
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 472
  • 472
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Western blot:
  • Yes
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Related Information

Frequently Bought Together