Anti-ITGAX Mouse Monoclonal Antibody (DyLight® 488) [clone: 3.9]
NOVUNB100-2711G
- Antibody type:Primary
- Antigen name:Integrin Subunit Alpha X
- Antigen symbol:ITGAX
- Clonality:Monoclonal
- Clone:3.9
- Conjugation:DyLight® 488
- Flow cytometry:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human
- Gene ID:3687
- Antigen synonyms:CD11C|SLEB6
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Type: Primary
Antigen: ITGAX (integrin, alpha X (complement component 3 receptor 4 subunit))
Clonality: monoclonal
Clone: 3.9
Conjugation: DyLight® 488
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: