Anti-JARID2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # NOVUNB100-2214H
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Jumonji AT Rich Interactive Domain 2
- Antigen symbol:JARID2
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Gene ID:3720
- Antigen synonyms:JMJ|JARID2|Protein Jumonji|Jumonji/ARID domain-containing protein 2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: JARID2 (jumonji, AT rich interactive domain 2)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: