Order Entry
Italy
ContactUsLinkComponent
 
Anti-JARID2 Rabbit Polyclonal Antibody (DyLight® 650)
  NOVUNB100-2214C
Anti-JARID2 Rabbit Polyclonal Antibody (DyLight® 650)
  NOVUNB100-2214C

 

  • Antibody type:
    Primary
  • Antigen name:
    Jumonji AT Rich Interactive Domain 2
  • Antigen symbol:
    JARID2
  • Clonality:
    Polyclonal
  • Conjugation:
    DyLight® 650
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Gene ID:
    3720
  • Antigen synonyms:
    JMJ|JARID2|Protein Jumonji|Jumonji/ARID domain-containing protein 2
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µL
  • Pk:
    0,1 mL

 

 

Type: Primary
Antigen: JARID2 (jumonji, AT rich interactive domain 2)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Rabbit
Isotype: none
Reactivity: