Anti-LSD1 Mouse Monoclonal Antibody [clone: 1B2E5]
Mouse Monoclonal LSD1 Antibody (1B2E5). Tested Applications: Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Human, Mouse, Primate.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:200-1:1000
Type: Primary
Antigen: LSD1
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
- Catalog No:
- NOVUNB100-1762SS
- NOVUNB100-1762
- Antigen symbol:
- KDM1A
- LSD1
- Antigen name:
- lysine (K)-specific demethylase 1A
- Lysine-specific histone demethylase 1A
- Antigen synonyms:
- BHC110|LSD1|KDM1|AOF2
- BHC110|BRAF35-HDAC complex protein BHC110|Flavin-containing amine oxidase domain-containing protein 2|LSD1|KDM1|Lysine-specific histone demethylase 1A|AOF2|RP1-184J9.1|KIAA0601
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- 1B2E5
- 1B2E5
- Reactivity:
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 23028
- 23028
- Isotype:
- IgG1
- IgG1
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ChIP:
- Yes
- Yes
- Storage buffer:
- Ascites
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Frequently Bought Together