Anti-ALDH3A1 Rabbit Polyclonal Antibody
Catalog # BSBTPA2074
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Aldehyde Dehydrogenase 3 Family, member A1
- Antigen symbol:ALDH3A1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-Terminal
- Cross adsorption:No
- Form:Lyophilized
- Amino acid number:440 - 453
- Sequence:KVRYPPSPAKMTQH
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human ALDH3A1 (440-453aa KVRYPPSPAKMTQH)
- Purification:Purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Rabbit IgG polyclonal antibody for Aldehyde dehydrogenase, dimeric NADP-preferring(ALDH3A1) detection. Tested with WB, IHC-P in Human.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: ALDH3A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human