Order Entry
Italy
ContactUsLinkComponent
Anti-ATF4 Rabbit Polyclonal Antibody
Anti-ATF4 Rabbit Polyclonal Antibody
Catalog # BSBTPA1537
Supplier:  Boster Bio
undefined
Anti-ATF4 Rabbit Polyclonal Antibody
Catalog # BSBTPA1537
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Activating transcription factor 4
  • Antigen symbol:
    ATF4
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    C-Terminal
  • Cross adsorption:
    No
  • Form:
    Lyophilized
  • Antigen synonyms:
    DNA-binding protein TAXREB67|cAMP-dependent transcription factor ATF-4|Cyclic AMP-dependent transcription factor ATF-4|TXREB|TaxREB67|Tax-responsive enhancer element-binding protein 67|CREB2|Activating transcription factor 4|cAMP-responsive element-binding protein 2|CREB-2|Cyclic AMP-responsive element-binding protein 2|ATF4
  • Amino acid number:
    332 - 351
  • Sequence:
    QYLKDLIEEVRKARGKKRVP
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ice
  • Immunogen:
    A synthetic peptide corresponding to a sequence at the C-terminus of human ATF4 (332-351aa QYLKDLIEEVRKARGKKRVP), identical to the related rat and mouse sequences.
  • Purification:
    purified by immunogen affinity purification
  • Size:
    100 µg
  • Pk:
    0,1 mg

Specifications

About this item

Rabbit IgG polyclonal antibody for Cyclic AMP-dependent transcription factor ATF-4(ATF4) detection. Tested with WB in Human; Mouse; Rat.

  • Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: ATF4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat