Anti-Caveolin-1 Rabbit Polyclonal Antibody
Catalog # BSBTPA1514
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Caveolin 1
- Antigen symbol:CAV1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:C-Terminal
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:CAV|VIP21|CAV1|MSTP085|caveolae protein 22kD|cell growth-inhibiting protein 32|caveolin 1
- Amino acid number:164 - 178
- Sequence:GKIFSNVRINLQKEI
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human Caveolin-1 (164-178aa GKIFSNVRINLQKEI), different from the related rat and mouse sequences by three amino acids.
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Rabbit IgG polyclonal antibody for Caveolin-1(CAV1) detection. Tested with WB, IHC-P, IHC-F, ICC in Human; Mouse; Rat.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: Caveolin-1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat