Anti-p53 Mouse Monoclonal Antibody [clone: IMD-53]
Catalog # BSBTMA1078
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Tumor protein p53
- Antigen symbol:P53
- Clonality:Monoclonal
- Clone:IMD-53
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Ascites
- Form:Lyophilised
- Antigen synonyms:or transformation-related protein 53 (TRP53)|tumor suppressor p53|antigen NY-CO-13|P53|BCC7|TRP53|phosphoprotein p53|LFS1|tumor protein p53
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:Recombinant human wild-type p53 protein
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Mouse IgG monoclonal antibody for p53, tumor protein p53 (TP53) detection. Tested with WB, IHC-P, IHC-F, ICC in Human. No cross reactivity with other proteins.
Reconstitute with 1,2% sodium acetate or neutral PBS.
If 1 ml of PBS is used, the antibody concentration will be 100 µg/ml.
Type: Primary
Antigen: p53
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human