Order Entry
Italy
ContactUsLinkComponent
 
Anti-IgG Rabbit Polyclonal Antibody (CF™ 680)
 :  Biotium
 
Anti-IgG Rabbit Polyclonal Antibody (CF™ 680)
 :  Biotium

 

  • Catalog No:
  • 20243-1
  • 20243.
  • Antigen symbol:
  • IgG
  • IgG
  • Antibody type:
  • Secondary
  • Secondary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • CF® 680
  • CF® 680
  • Reactivity:
  • Guinea pig
  • Guinea pig
  • Host:
  • Rabbit
  • Rabbit
  • Western blot:
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

 

 

Frequently Bought Together