Order Entry
Italy
ContactUsLinkComponent
Anti-SLD5 Rabbit Monoclonal Antibody [clone: EPR9318]
Anti-SLD5 Rabbit Monoclonal Antibody [clone: EPR9318]
Supplier:  Abcam
New Product
Certificates
undefined
Anti-SLD5 Rabbit Monoclonal Antibody [clone: EPR9318]
Supplier:  Abcam
New Product

Specifications

  • Catalog No:
  • ABCAAB139683-1ML
  • ABCAAB139683-100
  • Antigen symbol:
  • SLD5
  • Antibody type:
  • Primary
  • Clonality:
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Host:
  • Rabbit
  • Western blot:
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Flow cytometry:
  • Yes
  • Storage temperature:
  • Cool
  • Shipping temperature:
  • Ice

Specifications

Frequently Bought Together