Order Entry
Italy
ContactUsLinkComponent
Anti-TR4 Rabbit Polyclonal Antibody
Anti-TR4 Rabbit Polyclonal Antibody
  ABCAAB137582-50
 :  Abcam
undefined
Anti-TR4 Rabbit Polyclonal Antibody
  ABCAAB137582-50
 :  Abcam

 

  • Antibody type:
    Primary
  • Antigen name:
    nuclear receptor subfamily 2, group C, member 2
  • Antigen symbol:
    NR2C2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Seq:
    1202
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    50 µl
  • Pack type:
    Vial
  • Pk:
    50 µl

 

 

 :

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: NR2C2 (nuclear receptor subfamily 2, group C, member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human