Order Entry
Italy
ContactUsLinkComponent
 
Anti-TRAP/CD40L Mouse Monoclonal Antibody [clone: 24-31] (PE (Phycoerythrin))
  ABCAAB134405-50
 :  Abcam
undefined
Anti-TRAP/CD40L Mouse Monoclonal Antibody [clone: 24-31] (PE (Phycoerythrin))
  ABCAAB134405-50
 :  Abcam

 

  • Antibody type:
    Primary
  • Antigen name:
    CD40 ligand
  • Antigen symbol:
    CD40LG
  • Clonality:
    Monoclonal
  • Clone:
    24-31
  • Conjugation:
    PE (Phycoerythrin)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Seq:
    0780
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    50 Tests
  • Pack type:
    Vial
  • Pk:
    50 Tests

 

 

PE Mouse monoclonal [24-31] to TRAP/CD40L.

 :

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: TRAP/CD40L
Clonality: Monoclonal
Clone: 24-31
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human