Order Entry
Italy
ContactUsLinkComponent
 
Anti-Lambda Light chain Rabbit Monoclonal Antibody [clone: EPR5367-62]
 :  Abcam
New Product
 
undefined
Anti-Lambda Light chain Rabbit Monoclonal Antibody [clone: EPR5367-62]
 :  Abcam
New Product

 

  • Catalog No:
  • ABCAAB124719-1ML
  • ABCAAB124719-100
  • Antigen symbol:
  • IgG lambda
  • Antibody type:
  • Primary
  • Clonality:
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Clone:
  • EPR5367
  • Host:
  • Rabbit
  • Isotype:
  • IgG
  • Western blot:
  • Yes
  • ELISA:
  • Yes
  • ImmunoChemistry:
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Storage temperature:
  • Cool
  • Shipping temperature:
  • Ice

 

Frequently Bought Together